4-Methylumbelliferyl-2-acetamido-2-deoxy-6-sulphate-Β-D-glucopyranoside sodium salt

Lieferant: Apollo Scientific
Achtung

BIM1189-100MG BIM1189-25MG BIM1189-50MG
APOSBIM1189-100MGEA 638 EUR
APOSBIM1189-100MG APOSBIM1189-25MG APOSBIM1189-50MG
4-Methylumbelliferyl-2-acetamido-2-deoxy-6-sulphate-Β-D-glucopyranoside sodium salt
4-Methylumbelliferyl-2-acetamido-2-deoxy-6-sulphate-Β-D-glucopyranoside sodium salt

Fluorescent substrate for isoenzyme A of N-acetyl-b-D-glucosaminidase (Hexosaminidase A)

Molecular Weight: 481,50 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00132972
CAS-Nummer: 142439-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR