4-Nitrophenyl-α-D-mannopyranosid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Nitrophenyl α-D-Mannopyranoside, 4-Nitrophenyl a-D-mannopyranoside
BIM1208-1G
BIM1208-500MG
APOSBIM1208-1GEA
59.6
EUR
APOSBIM1208-1G
APOSBIM1208-500MG
4-Nitrophenyl-α-D-mannopyranosid
4-Nitrophenyl-α-D-mannopyranosid
Formel:
C₁₂H₁₅NO₈ Schmelzpunkt: 180 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00066002 CAS-Nummer: 10357-27-4 EINECS: 233-776-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...