Zeaxanthin
Lieferant: Apollo Scientific
Synonyme:
(1R,1'R)-4,4'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(3,5,5-trimethylcyclohex-3-enol)
BIM1908-10MG
BIM1908-25MG
BIM1908-50MG
APOSBIM1908-10MGEA
145
EUR
APOSBIM1908-10MG
APOSBIM1908-25MG
APOSBIM1908-50MG
Zeaxanthin
Zeaxanthin
Formel:
C₄₀H₅₆O₂ Molecular Weight: 568,89 g/mol Schmelzpunkt: 205…206 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD08435940 CAS-Nummer: 144-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...