4-Methylumbelliferyl phosphate freie Säure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Phosphoric Acid 4-Methylumbelliferyl Ester, 4-Methylumbelliferyl phosphoric acid, 4-Methyl-2-oxo-2H-chromen-7-yl dihydrogen phosphate
BIM2047-1G
BIM2047-5G
APOSBIM2047-1GEA
58
EUR
APOSBIM2047-1G
APOSBIM2047-5G
4-Methylumbelliferyl phosphate freie Säure 98%
4-Methylumbelliferyl phosphate
Formel:
C₁₀H₉O₆P Molecular Weight: 256,15 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00016969 CAS-Nummer: 3368-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...