Methylthymolblau Natriumsalz

Lieferant: Apollo Scientific

Synonyme: Methylthymolblau-Komplexon, Methylthymolblau Tetranatriumsalz, Methyl thymol blue complexone

BIM5000-1G BIM5000-5G BIM5000-25G BIM5000-100G
APOSBIM5000-1GEA 21.9 EUR
APOSBIM5000-1G APOSBIM5000-5G APOSBIM5000-25G APOSBIM5000-100G
Methylthymolblau Natriumsalz
Methylthymolblau Natriumsalz

Indicator for metal titrations

Formel: C₃₇H₄₀N₂Na₄O₁₃S
Molecular Weight: 844,76 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00148905
CAS-Nummer: 1945-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR