Biotin-C5-Amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Biotinamido)pentylamine, N-(5-Aminopentyl)biotinamide, N-(5-Aminopentyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide
BIMB102-100MG
BIMB102-25MG
BIMB102-50MG
APOSBIMB102-100MGEA
0
EUR
APOSBIMB102-100MG
APOSBIMB102-25MG
APOSBIMB102-50MG
Biotin-C5-Amine
Biotin-C5-Amine
Formel:
C₁₅H₂₈N₄O₂S Molecular Weight: 328,48 g/mol Schmelzpunkt: 132…135 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00216002 CAS-Nummer: 115416-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...