X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA) ≥98%
Lieferant: Apollo Scientific
Achtung
Synonyme: Cyclohexylammoniumsalz der 5-Brom-4-chlor-3-indolyl-β-D-glucuronsäure, X-Glucuro CHAsalz, Cyclohexylammoniumsalz der (2S,3S,4S,5R)-6-[(5-Brom-4-chlor-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxan-2-carbonsäure
BIMB1021-1G BIMB1021-250MG BIMB1021-500MG
APOSBIMB1021-1GEA 114 EUR
APOSBIMB1021-1G APOSBIMB1021-250MG APOSBIMB1021-500MG
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA) ≥98%
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA)
X-GlcA is a substrate for b-Glucuronidase (GUS) encoded by the gusA gene. The substrate is used as a qualitative marker of specific GUS expression.
Formel: C₂₀H₂₆BrClN₂O₇
Molecular Weight: 521.79 g/mol
Schmelzpunkt: 230
Lagertemperatur: Kühlschrank
MDL: MFCD00058543
CAS-Nummer: 114162-64-0