6-Chloro-3-indolyl-β-D-galactopyranoside ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Rose-Gal, Salmon-Gal
BIMB1026-100MG
BIMB1026-1G
BIMB1026-5G
APOSBIMB1026-100MGEA
37.7
EUR
APOSBIMB1026-100MG
APOSBIMB1026-1G
APOSBIMB1026-5G
6-Chloro-3-indolyl-β-D-galactopyranoside ≥98%
6-Chloro-3-indolyl-beta-D-galactopyranoside
Salmon-Gal is an alternative chromogenic substrate for b-D-Galactisidase. Salmon-Gal is used in conjunctionwith IPTG for detection of b-Galactosidase activityin bacterial colonies.
Formel:
C₁₄H₁₆ClNO₆ Molecular Weight: 329.733 g/mol Schmelzpunkt: 188…190 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00467206 CAS-Nummer: 138182-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...