5-Bromo-3-indolyl-β-D-galactopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: Bluo-Gal, (2S,3R,4S,5R,6R)-2-((5-Bromo-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, 5-Bromo-3-indolyl β-D-galactopyranoside

BIMB1030-100MG BIMB1030-1G BIMB1030-5G
APOSBIMB1030-100MGEA 21.9 EUR
APOSBIMB1030-100MG APOSBIMB1030-1G APOSBIMB1030-5G
5-Bromo-3-indolyl-β-D-galactopyranoside
5-Bromo-3-indolyl-beta-D-galactopyranoside

Chromogenic substrate for b-Galactosidase. Reported to produce a deeper blue water insoluble colouration than X-Gal.

Formel: C₁₄H₁₆BrNO₆
Molecular Weight: 374,19 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00063691
CAS-Nummer: 97753-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR