3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS) ≥99,0%, ultrarein
Lieferant: Apollo Scientific
Achtung
Synonyme: CHAPS
BIMB1085-10G BIMB1085-25G BIMB1085-50G
APOSBIMB1085-10GEA 50.9 EUR
APOSBIMB1085-10G APOSBIMB1085-25G APOSBIMB1085-50G
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS) ≥99,0%, ultrarein
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
A nondenaturing zwitterionic detergent for membrane chemistry.
  • Purity (HPLC) >99.0%
  • Purity (TLC) >99.0%
  • Abs. (10% water) @ 260nm <0.1
Formel: C₃₂H₅₈N₂O₇S
Molecular Weight: 614.89 g/mol
Schmelzpunkt: 156…158 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00081083
CAS-Nummer: 75621-03-3
Merck Index: 13,02052
Bestellen
TECHNISCHE DATEN
Environmentally Preferable