3-Indolyl-ß-D-galactopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: Indol-3-yl-β-D-galactopyranoside

BIMB1284-250MG BIMB1284-500MG BIMB1284-1G
APOSBIMB1284-250MGEA 69.6 EUR
APOSBIMB1284-250MG APOSBIMB1284-500MG APOSBIMB1284-1G
3-Indolyl-ß-D-galactopyranoside
3-Indolyl-ß-D-galactopyranoside
Formel: C₁₄H₁₇NO₆
Molecular Weight: 295,291 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00083389
CAS-Nummer: 126787-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR