5-Bromo-4-chloro-3-indolyl-α-D-galactoside ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
X-a-Gal, 5-Bromo-4-Chloro-3-Indolyl-a-D Galactopyranoside, X-alpha-Gal
BIMB1301-100MG
BIMB1301-1G
BIMB1301-250MG
APOSBIMB1301-100MGEA
65.4
EUR
APOSBIMB1301-100MG
APOSBIMB1301-1G
APOSBIMB1301-250MG
5-Bromo-4-chloro-3-indolyl-α-D-galactoside ≥98%
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
X-a-Gal is a chromogenic substrate which can beused to demonstrate a-Galactosidase activity.
Formel:
C₁₄H₁₅BrClNO₆ Molecular Weight: 408,63 g/mol Schmelzpunkt: 170…173 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063780 CAS-Nummer: 107021-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...