Leupeptin trifluoroacetate salt ≥90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Acetyl-Leu-Leu-Arg-al trifluoroacetate salt
BIMI1322-10MG
BIMI1322-5MG
APOSBIMI1322-10MGEA
87
EUR
APOSBIMI1322-10MG
APOSBIMI1322-5MG
Leupeptin trifluoroacetate salt ≥90%
Leupeptin trifluoroacetate salt
Inhibitor of serine and cysteine proteases.
Formel:
C₂₀H₃₈N₆O₄·xC₂HF₃O₂ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00133458 CAS-Nummer: 147385-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...