2'-Amino-2'-deoxyinosine

Lieferant: Apollo Scientific

BIMN2007-100MG BIMN2007-250MG BIMN2007-500MG
APOSBIMN2007-100MGEA 306 EUR
APOSBIMN2007-100MG APOSBIMN2007-250MG APOSBIMN2007-500MG
2'-Amino-2'-deoxyinosine
2'-Amino-2'-deoxyinosine
Formel: C₁₀H₁₃N₅O₄
Lagertemperatur: Kühlschrank
MDL: MFCD09750831
CAS-Nummer: 75763-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR