2'-Fluoro-2'-deoxyinosine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2’-Deoxy-2’-fluoroinosine, 2'-Deoxy-2'-fluoroinosine, 9-((2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol

BIMN2011-100MG BIMN2011-1G BIMN2011-250MG
APOSBIMN2011-100MGEA 102 EUR
APOSBIMN2011-100MG APOSBIMN2011-1G APOSBIMN2011-250MG
2'-Fluoro-2'-deoxyinosine
2'-Fluoro-2'-deoxyinosine
Formel: C₁₀H₁₁FN₄O₄
Molecular Weight: 270,22 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09750860
CAS-Nummer: 80049-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR