Nystatin Dihydrat

Lieferant: Apollo Scientific
Achtung

Synonyme: Fungicidin dihydrate, Mycostatin dihydrate

BIN0138-5G
APOSBIN0138-5GEA 94.4 EUR
APOSBIN0138-5G
Nystatin Dihydrat
Nystatin Dihydrat

Nystatin is a polyene antifungal antibiotic produced by streptomyces noursei.


  • Potency: 5210 IU/mg

Formel: C₄₇H₇₅NO₁₇·2H₂O
Molecular Weight: 926,09 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00036240
CAS-Nummer: 1400-61-9
EINECS: 215-749-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR