1-Naphthylessigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: α-Naphthylessigsäure, Planofix, 1-Naphthalinessigsäure

BIN0600-100G BIN0600-500G BIN0600-1KG
APOSBIN0600-100GEA 31.9 EUR
APOSBIN0600-100G APOSBIN0600-500G APOSBIN0600-1KG
1-Naphthylessigsäure
1-Naphthylessigsäure

NAA is a synthetic plant hormone belonging to the Auxin family, with numerous applications including plant propagation and plant cell culture. It can be added to plant media such as Murashige and Skoog and Chu N6 to help induce root formation..

Formel: C₁₂H₁₀O₂
Molecular Weight: 186.21 g/mol
Siedepunkt: 352
Schmelzpunkt: 129…132 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00004046
CAS-Nummer: 86-87-3
EINECS: 201-705-8
Merck Index: 12,06458

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR