Nonyl-β-D-glucopyranosid

Lieferant: Apollo Scientific
Achtung

Synonyme: Nonyl-β-D-glucosid, NG

BIN5001-500MG BIN5001-1G BIN5001-5G
APOSBIN5001-500MGEA 97.3 EUR
APOSBIN5001-500MG APOSBIN5001-1G APOSBIN5001-5G
Nonyl-β-D-glucopyranosid
Nonyl-β-D-glucopyranosid
Formel: C₁₅H₃₀O₆
Molecular Weight: 306,4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00063300
CAS-Nummer: 69984-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR