Parthenolide

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,5alpha-Epoxy-6beta-hydroxygermacra-1(10),11(13)-dien-12-oic Acid gamma-Lactone, (3aS,9aR,10aR,10bS,E)-6,9a-Dimethyl-3-methylene-3a,4,5,8,9,9a,10a,10b-octahydrooxireno[2',3':9,10]cyclodeca[1,2-b]furan-2(3H)-one

BIP0110-100MG BIP0110-250MG BIP0110-50MG
APOSBIP0110-100MGEA 66.7 EUR
APOSBIP0110-100MG APOSBIP0110-250MG APOSBIP0110-50MG
Parthenolide
Parthenolide

Anti-inflammatory agent

Formel: C₁₅H₂₀O₃
Molecular Weight: 248,32 g/mol
Schmelzpunkt: 115…116 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00134592
CAS-Nummer: 20554-84-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR