PX-866

Lieferant: Apollo Scientific
Achtung

BIP1009-10MG BIP1009-5MG
APOSBIP1009-10MGEA 275 EUR
APOSBIP1009-10MG APOSBIP1009-5MG
PX-866
PX-866

PX-866 is a synthetic derivate of wortmannin

Lagertemperatur: Gefrierschrank
CAS-Nummer: 502632-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR