6-Benzylaminopurin-Lösung 1,0 mg/ml
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzyl(purin-6-yl)amin, 6-Benzyladenin, N6-Benzyladenin
BIPB130-100ML
BIPB130-500ML
APOSBIPB130-100MLEA
50.9
EUR
APOSBIPB130-100ML
APOSBIPB130-500ML
6-Benzylaminopurin-Lösung 1,0 mg/ml
6-Benzylaminopurin
- Plant tissue culture tested
Formel:
C₁₂H₁₁N₅ Molecular Weight: 225,25 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00005572 CAS-Nummer: 1214-39-7 EINECS: 214-927-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...