Amino-PEG4-acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Amino-PEG4-acid, Amino-PEG-acid, α-Amine-ω-propionic acid tetraethylene glycol, 3-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]propionic acid, NH2-PEG4-COOH, Carboxy-PEG4-amine, Tetraethylene glycol amino propionic acid, NH2-PEG4-CH2CH2COOH, CA(PEG)4, (PEO)4-Aminopropionic acid, 1-Amino-3,6,9,12-tetraoxapentadecan-15-oic acid, Tetraethylene glycol α-amino-ω-propionic acid
BIPG1033-100MG
BIPG1033-250MG
BIPG1033-500MG
APOSBIPG1033-100MGEA
139
EUR
APOSBIPG1033-100MG
APOSBIPG1033-250MG
APOSBIPG1033-500MG
Amino-PEG4-acid
Amino-PEG4-acid
Formel:
C₁₁H₂₃NO₆ Molecular Weight: 265.30 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11041129 CAS-Nummer: 663921-15-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...