Amino-PEG6-acid

Lieferant: Apollo Scientific
Achtung

BIPG1035-100MG BIPG1035-50MG BIPG1035-250MG
APOSBIPG1035-100MGEA 174 EUR
APOSBIPG1035-100MG APOSBIPG1035-50MG APOSBIPG1035-250MG
Amino-PEG6-acid
Amino-PEG6-acid
Formel: C₁₅H₃₁NO₈
Molecular Weight: 353.41 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 905954-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR