Azido-PEG4-acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
15-Azido-4,7,10,13-tetraoxapentadecanoic acid
BIPG1106-250MG
BIPG1106-50MG
BIPG1106-100MG
APOSBIPG1106-250MGEA
345
EUR
APOSBIPG1106-250MG
APOSBIPG1106-50MG
APOSBIPG1106-100MG
Azido-PEG4-acid
Azido-PEG4-acid
Formel:
C₁₁H₂₁N₃O₆ Molecular Weight: 291,3 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD12406155 CAS-Nummer: 1257063-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...