Azido-PEG24-alcohol

Lieferant: Apollo Scientific
Achtung

BIPG1120-100MG BIPG1120-250MG
APOSBIPG1120-100MGEA 364 EUR
APOSBIPG1120-100MG APOSBIPG1120-250MG
Azido-PEG24-alcohol
Azido-PEG24-alcohol
Formel: C₄₈H₉₇N₃O₂₄
Molecular Weight: 1100,29 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 73342-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR