Biotin-PEG3-Amine

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethyl]biotinamide, N-(2-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, N-Biotinyl-3,6,9-trioxaundecane-1,11-diamine, Amine-PEG3-Biotin

BIPG1208-100MG BIPG1208-50MG
APOSBIPG1208-100MGEA 161 EUR
APOSBIPG1208-100MG APOSBIPG1208-50MG
Biotin-PEG3-Amine
Biotin-PEG3-amine
Formel: C₁₈H₃₄N₄O₅S
Molecular Weight: 418,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09952640
CAS-Nummer: 359860-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR