Fmoc-N-amido-dPEG2-acid

Lieferant: Apollo Scientific

BIPG1396-500MG BIPG1396-1G BIPG1396-100MG
APOSBIPG1396-500MGEA 145 EUR
APOSBIPG1396-500MG APOSBIPG1396-1G APOSBIPG1396-100MG
Fmoc-N-amido-dPEG2-acid
Fmoc-N-amido-dPEG2-acid
Formel: C₂₂H₂₅NO₆
Molecular Weight: 399.44 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 872679-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR