Mal-amido-PEG2-acid

Lieferant: Apollo Scientific

Synonyme: 3-(2-(2-(3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)ethoxy)ethoxy)propanoic acid, Mal-amido-PEG2-C2-acid

BIPG1472-1G BIPG1472-250MG
APOSBIPG1472-1GEA 310 EUR
APOSBIPG1472-1G APOSBIPG1472-250MG
Mal-amido-PEG2-acid
Mal-amido-PEG2-acid
Formel: C₁₄H₂₀N₂O₇
Molecular Weight: 328.32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11041136
CAS-Nummer: 756525-98-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR