Undecaethylene glycol

Lieferant: Apollo Scientific

Synonyme: 3,6,9,12,15,18,21,24,27,30-Decaoxadotriacontane-1,32-diol, PEG-11, HO-PEG11-OH

BIPG1677-1G BIPG1677-100MG BIPG1677-500MG
APOSBIPG1677-1GEA 348 EUR
APOSBIPG1677-1G APOSBIPG1677-100MG APOSBIPG1677-500MG
Undecaethylene glycol
Undecaethylene glycol
Formel: C₂₂H₄₆O₁₂
Molecular Weight: 502,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00698693
CAS-Nummer: 5579-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR