Thiol-PEG4-acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Mercapto-3,6,9,12-tetraoxapentadecan-15-oic acid
BIPG1838-250MG
APOSBIPG1838-250MGEA
270
EUR
APOSBIPG1838-250MG
Thiol-PEG4-acid
Thiol-PEG4-acid
Formel:
C₁₁H₂₂O₆S Molecular Weight: 282,35 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
749247-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...