Streptomycinsulfat
Lieferant: Apollo Scientific
Achtung
BIS0148-100G BIS0148-25G
APOSBIS0148-100GEA 60.9 EUR
APOSBIS0148-100G APOSBIS0148-25G
Streptomycinsulfat
Streptomycin sulphate
Streptomycin is an aminoglycoside antibiotic and has a bactericidal action against many gram-negative bacteria.
  • Cell culture tested
Formel: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Molecular Weight: 1457.4 g/mol
Schmelzpunkt: 250
Lagertemperatur: Kühlschrank
MDL: MFCD00037023
CAS-Nummer: 3810-74-0
EINECS: 223-286-0
Merck Index: 12,08983