SB203580

Lieferant: Apollo Scientific
Gefahr

Synonyme: SB 203580, 4-(5-(4-Fluorophenyl)-2-(4-(methylsulphinyl)phenyl)-1H-imidazol-4-yl)pyridine

BIS0733-100MG BIS0733-25MG BIS0733-50MG
APOSBIS0733-100MGEA 277 EUR
APOSBIS0733-100MG APOSBIS0733-25MG APOSBIS0733-50MG
SB203580
SB203580

Selective inhibitor of p38 MAP kinase. Inhibitor of p40 kinase .

Formel: C₂₁H₁₆FN₃OS
Molecular Weight: 377,4 g/mol
Schmelzpunkt: 240…243 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00922198
CAS-Nummer: 152121-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR