1,9-Pyrazoloanthrone ≥99%
Lieferant: Apollo Scientific
Achtung
Synonyme: JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125
BIS1100-1G BIS1100-300MG
APOSBIS1100-1GEA 444 EUR
APOSBIS1100-1G APOSBIS1100-300MG
1,9-Pyrazoloanthrone ≥99%
1,9-Pyrazoloanthrone
Novel, potent and selective JNK-1,-2, and -3 inhibitor (Ki = 0.19 uM). SP600125 is an ATP-competitive reversible inhibitor with >20-fold selectivity vs. a range of kinases and enzymes tested. Proc. Natl. Acad. Sci. USA 98: 13681-13686 (2001).
Formel: C₁₄H₈N₂O
Molecular Weight: 220,231 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00022289
CAS-Nummer: 129-56-6