Scopoletin (7-Hydroxy-6-methoxy-2H-chromen-2-on)

Lieferant: Apollo Scientific
Achtung

Synonyme: Scopoletin, 7-Hydroxy-6-methoxy-2H-chromen-2-one

BIS1402-100MG BIS1402-500MG BIS1402-1G
APOSBIS1402-100MGEA 50.9 EUR
APOSBIS1402-100MG APOSBIS1402-500MG APOSBIS1402-1G
Scopoletin (7-Hydroxy-6-methoxy-2H-chromen-2-on)
Scopoletin (7-Hydroxy-6-methoxycumarin)

Dye to detect active oxygen species during the oxidative burst.

Formel: C₁₀H₈O₄
Molecular Weight: 192.1681 g/mol
Schmelzpunkt: 203…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006872
CAS-Nummer: 92-61-5
EINECS: 202-177-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR