Streptozocin

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Methyl-1-nitroso-3-((2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)urea, Streptozotocin

BIS4356-1G BIS4356-250MG BIS4356-500MG
APOSBIS4356-1GEA 702 EUR
APOSBIS4356-1G APOSBIS4356-250MG APOSBIS4356-500MG
Streptozocin
Streptozocin

Potent methylating agent for DNA

Formel: C₈H₁₅N₃O₇
Molecular Weight: 265,2 g/mol
Schmelzpunkt: 121 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00006607
CAS-Nummer: 18883-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR