(S)-O-Desmethylraclopride

Lieferant: Apollo Scientific
Gefahr

Synonyme: (S)-3,5-Dichloro-N-((1-ethylpyrrolidin-2-yl)methyl)-2,6-dihydroxybenzamide, (S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide

BISC1012-25MG BISC1012-5MG
APOSBISC1012-25MGEA 0 EUR
APOSBISC1012-25MG APOSBISC1012-5MG
(S)-O-Desmethylraclopride
(S)-O-Desmethylraclopride
Formel: C₁₄H₁₈Cl₂N₂O₃
Molecular Weight: 333,21 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD02179385
CAS-Nummer: 119670-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR