LY-2157299
Lieferant: Apollo Scientific
Synonyme:
4-(2-(6-Methylpyridin-2-yl)-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazol-3-yl)quinoline-6-carboxamide, Galunisertib
BISN0107-25MG
BISN0107-100MG
APOSBISN0107-25MGEA
39.3
EUR
APOSBISN0107-25MG
APOSBISN0107-100MG
LY-2157299
LY-2157299
LY2157299 is a potent TGFβ receptor I (TβRI) inhibitor with IC50 of 56 nM in a cell-free assay.
Formel:
C₂₂H₁₉N₅O Molecular Weight: 369,42 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12923319 CAS-Nummer: 700874-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...