TAK-242

Lieferant: Apollo Scientific

Synonyme: Resatorvid, (R)-Ethyl 6-(N-(2-chloro-4-fluorophenyl)sulphamoyl)cyclohex-1-enecarboxylate, 6R-[[(2-chloro-4-fluorophenyl)amino]sulphonyl]-1-cyclohexene-1-carboxylic acid, ethyl ester

BISN0117-5MG BISN0117-10MG
APOSBISN0117-5MGEA 122 EUR
APOSBISN0117-5MG APOSBISN0117-10MG
TAK-242
TAK-242

TAK-242, a small-molecule-specific inhibitor of Toll-like receptor (TLR)4 signaling, inhibits the production of lipopolysaccharide-induced inflammatory mediators by binding to the intracellular domain of TLR4.

Formel: C₁₅H₁₇NO₄ClFS
Molecular Weight: 361,8 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11046362
CAS-Nummer: 243984-11-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR