Bortezomib
Lieferant: Apollo Scientific
BISN0125-5MG
BISN0125-10MG
BISN0125-50MG
APOSBISN0125-5MGEA
109
EUR
APOSBISN0125-5MG
APOSBISN0125-10MG
APOSBISN0125-50MG
Bortezomib
Bortezomib
Bortezomib is an inhibitor of 20S proteasome with Ki of 0.6 nM.
Formel:
C₁₉H₂₅BN₄O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09056737 CAS-Nummer: 179324-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...