BAY 11-7082

Lieferant: Apollo Scientific

BISN0126-10MG BISN0126-50MG
APOSBISN0126-10MGEA 85.9 EUR
APOSBISN0126-10MG APOSBISN0126-50MG
BAY 11-7082
BAY 11-7082

BAY 11-7082 is a NF-B inhibitor, inhibits TNF-induced IB phosphorylation with IC50 of 10 M in tumor cells. It also inhibits components of the ubiquitin system through inactivating the E2-conjugating enzymes Ubc (ubiquitin conjugating) 13 and UbcH7 and the E3 ligase LUBAC (linear ubiquitin assembly complex).

Formel: C₁₀H₉NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00712162
CAS-Nummer: 19542-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR