Y-27632 dihydrochloride
Lieferant: Apollo Scientific
BISN0135-10MG
BISN0135-100MG
APOSBISN0135-10MGEA
116
EUR
APOSBISN0135-10MG
APOSBISN0135-100MG
Y-27632 dihydrochloride
Y-27632 dihydrochloride
Y-27632 dihydrochloride is a selective ROCK1 p160ROCK inhibitor with Ki of 140 nM exhibits 200-fold selectivity over other kinases including PKC cAMP-dependent protein kinase MLCK and PAK.
Formel:
C₁₄H₂₁N₃O·2HCl Lagertemperatur: Kühlschrank |
MDL:
MFCD03490488 CAS-Nummer: 129830-38-2 | 146986-50-7 (free base) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...