FK-866
Lieferant: Apollo Scientific
Synonyme:
(E)-N-(4-(1-Benzoylpiperidin-4-yl)butyl)-3-(pyridin-3-yl)acrylamide, (E)-Daporinad
BISN0147-1MG
BISN0147-5MG
BISN0147-10MG
APOSBISN0147-1MGEA
79.9
EUR
APOSBISN0147-1MG
APOSBISN0147-5MG
APOSBISN0147-10MG
FK-866
FK-866
FK866 effectively inhibits nicotinamide phosphoribosyltransferase (NMPRTase) with IC50 of 0.09 nM in a cell-free assay.
Formel:
C₂₄H₂₉N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10565943 CAS-Nummer: 658084-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...