MCC950 sodium salt
Lieferant: Apollo Scientific
Synonyme: MCC950, MCC950 sodium, Sodium ((1,2,3,5,6,7-hexahydro-s-indacen-4-yl)carbamoyl)((4-(2-hydroxypropan-2-yl)furan-2-yl)sulphonyl)amide
BISN0154-5MG BISN0154-10MG
APOSBISN0154-5MGEA 166 EUR
APOSBISN0154-5MG APOSBISN0154-10MG
MCC950 sodium salt
MCC950 sodium salt
MCC950 Sodium is a potent and highly specific small molecule inhibitor of NLRP3 inflammasome with IC50 of 7.5 nM in BMDMs.
Formel: C₂₀H₂₃N₂NaO₅S
Molecular Weight: 426,5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30478884
CAS-Nummer: 256373-96-3