GDC-0449
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-N-(4-chloro-3-(pyridin-2-yl)phenyl)-4-(methylsulphonyl)benzamide, Vismodegib
BISN0157-10MG
BISN0157-50MG
BISN0157-100MG
APOSBISN0157-10MGEA
21.9
EUR
APOSBISN0157-10MG
APOSBISN0157-50MG
APOSBISN0157-100MG
GDC-0449
GDC-0449
Vismodegib is an inhibitor of hedgehog pathway with IC50 of 3 nM and also inhibits P-gp IC50 30 M.
Formel:
C₁₉H₁₄Cl₂N₂O₃S Molecular Weight: 421,3 g/mol Schmelzpunkt: 180…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12407408 CAS-Nummer: 879085-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...