Sonidegib
Lieferant: Apollo Scientific
Synonyme:
N-(6-(cis-2,6-Dimethylmorpholino)pyridin-3-yl)-2-methyl-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-carboxamide, LDE225
BISN0158-5MG
BISN0158-10MG
BISN0158-50MG
APOSBISN0158-5MGEA
79.9
EUR
APOSBISN0158-5MG
APOSBISN0158-10MG
APOSBISN0158-50MG
Sonidegib
Sonidegib
Erismodegib, NVP-LDE225) is a Smoothened (Smo) antagonist, inhibiting Hedgehog (Hh) signaling with IC50 of 1.3 nM (mouse) and 2.5 nM (human) in cell-free assays, respectively.
Formel:
C₂₆H₂₆F₃N₃O₃ Molecular Weight: 485.5 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16038928 CAS-Nummer: 956697-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...