Doramapimod

Lieferant: Apollo Scientific

Synonyme: p38 MAP Kinase Inhibitor X, 1-(3-(tert-Butyl)-1-(p-tolyl)-1H-pyrazol-5-yl)-3-(4-(2-morpholinoethoxy)naphthalen-1-yl)urea, BIRB 796

BISN0161-5MG BISN0161-10MG
APOSBISN0161-5MGEA 31.9 EUR
APOSBISN0161-5MG APOSBISN0161-10MG
Doramapimod
Doramapimod

MAPK/ERK Signalling Pathway

Formel: C₃₁H₃₇N₅O₃
Molecular Weight: 527,669 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09752957
CAS-Nummer: 285983-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR