PD 169316

Lieferant: Apollo Scientific

Synonyme: 4-(4-(4-Fluorophenyl)-2-(4-nitrophenyl)-1H-imidazol-5-yl)pyridine, 4-[4-(4-fluorophenyl)-2-(4-nitrophenyl)-1H-imidazol-5-yl]pyridine

BISN0170-1MG BISN0170-5MG BISN0170-10MG
APOSBISN0170-1MGEA 58 EUR
APOSBISN0170-1MG APOSBISN0170-5MG APOSBISN0170-10MG
PD 169316
PD 169316

MAPK/ERK Signalling Pathway

Formel: C₂₀H₁₃FN₄O₂
Molecular Weight: 360,34 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12405019
CAS-Nummer: 152121-53-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR