PF-04708671

Lieferant: Apollo Scientific

Synonyme: PF-4708671, 2-((4-(5-Ethylpyrimidin-4-yl)piperazin-1-yl)methyl)-6-(trifluoromethyl)-1H-benzo[d]imidazole

BISN0176-5MG
APOSBISN0176-5MGEA 26.1 EUR
APOSBISN0176-5MG
PF-04708671
PF-04708671

MAPK/ERK Signalling Pathway

Formel: C₁₉H₂₁F₃N₆
Molecular Weight: 390,4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD18086922
CAS-Nummer: 1255517-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR