VX-745

Lieferant: Apollo Scientific

Synonyme: 5-(2,6-Dichlorophenyl)-2-((2,4-difluorophenyl)thio)-6H-pyrimido[1,6-b]pyridazin-6-one, Neflamapimod

BISN0180-5MG BISN0180-10MG BISN0180-50MG
APOSBISN0180-5MGEA 72.5 EUR
APOSBISN0180-5MG APOSBISN0180-10MG APOSBISN0180-50MG
VX-745
VX-745

MAPK/ERK Signalling Pathway

Formel: C₁₉H₉Cl₂F₂N₃OS
Molecular Weight: 436,3 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09834070
CAS-Nummer: 209410-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR