AS-703026
Lieferant: Apollo Scientific
Synonyme:
(S)-N-(2,3-Dihydroxypropyl)-3-((2-fluoro-4-iodophenyl)amino)isonicotinamide, MSC1936369B, Pimasertib
BISN0186-1MG
APOSBISN0186-1MGEA
29
EUR
APOSBISN0186-1MG
AS-703026
AS-703026
MAPK/ERK Signalling Pathway
Formel:
C₁₅H₁₅FIN₃O₃ Molecular Weight: 431,2 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD16660676 CAS-Nummer: 1236699-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...